5-Methyl-2-(trifluoromethoxy)benzenesulphonamide 97%
Leverantör: Apollo Scientific
PC303295-1G PC303295-250MG
APOSPC303295-1GEA 21450 SEK
APOSPC303295-1G APOSPC303295-250MG
5-Methyl-2-(trifluoromethoxy)benzenesulphonamide 97%
5-Methyl-2-(trifluoromethoxy)benzenesulphonamide
Formel: C₈H₈F₃NO₃S
MW: 255.214 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18399730
CAS nummer: 1261683-36-6