2-Fluoro-5-methyl-3-(trifluoromethyl)phenylacetic acid 98%
Leverantör: Apollo Scientific
PC303223-1G PC303223-250MG
APOSPC303223-1GEA 10600 SEK
APOSPC303223-1G APOSPC303223-250MG
2-Fluoro-5-methyl-3-(trifluoromethyl)phenylacetic acid 98%
2-Fluoro-5-methyl-3-(trifluoromethyl)phenylacetic acid
Formel: C₁₀H₈F₄O₂
MW: 236,163 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28054191
CAS nummer: 1706452-86-9