2,3-Difluoro-5-methoxy-4-methylbenzaldehyde 97%
Leverantör: Apollo Scientific
PC303212-1G
PC303212-250MG
APOSPC303212-1GEA
9975
SEK
APOSPC303212-1G
APOSPC303212-250MG
2,3-Difluoro-5-methoxy-4-methylbenzaldehyde 97%
2,3-Difluoro-5-methoxy-4-methylbenzaldehyde
Formel:
C₉H₈F₂O₂ MW: 186,155 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD27986838 CAS nummer: 1706430-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...