2-Chloro-3-(trifluoromethyl)benzenesulphonyl chloride 97%
Leverantör: Apollo Scientific
Fara
Synonyms: 2-Chloro-3-(chlorosulphonyl)benzotrifluoride
PC303179-1G PC303179-250MG
APOSPC303179-1GEA 5175 SEK
APOSPC303179-1G APOSPC303179-250MG
2-Chloro-3-(trifluoromethyl)benzenesulphonyl chloride 97%
2-Chloro-3-(trifluoromethyl)benzenesulphonyl chloride
Formel: C₇H₃Cl₂F₃O₂S
Storage Temperature: Omgivande
MDL Number: MFCD13185469
CAS nummer: 1214346-10-7
UN: 3265
ADR: 8,II