2-Fluoro-5-methyl-3-(trifluoromethyl)benzyl bromide 97%
Leverantör: Apollo Scientific
PC303170-1G PC303170-250MG
APOSPC303170-1GEA 7300 SEK
APOSPC303170-1G APOSPC303170-250MG
2-Fluoro-5-methyl-3-(trifluoromethyl)benzyl bromide 97%
2-Fluoro-5-methyl-3-(trifluoromethyl)benzyl bromide
Formel: C₉H₇BrF₄
MW: 271.049 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28054188
CAS nummer: 1706438-74-5