3-Chloro-5-(trifluoromethoxy)benzyl alcohol 97%

Leverantör: Apollo Scientific
Varning

PC302948-10G PC302948-5G
APOSPC302948-10GEA 5300 SEK
APOSPC302948-10G APOSPC302948-5G
3-Chloro-5-(trifluoromethoxy)benzyl alcohol 97%
3-Chloro-5-(trifluoromethoxy)benzyl alcohol
Formel: C₈H₆ClF₃O₂
MW: 226,58 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06660310
CAS nummer: 433926-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR