3-Chloro-4-ethoxy-5-fluorobenzyl bromide 98%
Leverantör: Apollo Scientific
PC302945-10G PC302945-5G PC302945-1G
APOSPC302945-10GEA 4775 SEK
APOSPC302945-10G APOSPC302945-5G APOSPC302945-1G
3-Chloro-4-ethoxy-5-fluorobenzyl bromide 98%
3-Chloro-4-ethoxy-5-fluorobenzyl bromide
Formel: C₉H₉BrClFO
MW: 267.523 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09832342
CAS nummer: 1017778-91-4