3-Chloro-2-fluoro-4-(trifluoromethyl)benzoic acid
Leverantör: Apollo Scientific
Varning
PC302830-10G PC302830-1G PC302830-5G
APOSPC302830-10GEA 11625 SEK
APOSPC302830-10G APOSPC302830-1G APOSPC302830-5G
3-Chloro-2-fluoro-4-(trifluoromethyl)benzoic acid
3-Chloro-2-fluoro-4-(trifluoromethyl)benzoic acid
Formel: C₈H₃ClF₄O₂
MW: 242.555 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD23795337
CAS nummer: 1431329-63-3