4-Chloro-3,5-difluorobenzoic acid 97%
Leverantör: Apollo Scientific
PC302575-5G PC302575-1G PC302575-25G
APOSPC302575-5GEA 2575 SEK
APOSPC302575-5G APOSPC302575-1G APOSPC302575-25G
4-Chloro-3,5-difluorobenzoic acid 97%
4-Chloro-3,5-difluorobenzoic acid
Formel: C₇H₃ClF₂O₂
MW: 192,547 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11554154
CAS nummer: 1160573-19-2