2-Methoxy-4-(trifluoromethyl)benzoyl chloride 97%
Leverantör: Apollo Scientific
Fara
PC302465-10G PC302465-5G PC302465-1G
APOSPC302465-10GEA 4725 SEK
APOSPC302465-10G APOSPC302465-5G APOSPC302465-1G
2-Methoxy-4-(trifluoromethyl)benzoyl chloride 97%
2-Methoxy-4-(trifluoromethyl)benzoyl chloride
Formel: C₉H₆ClF₃O₂
MW: 238,591 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04115977
CAS nummer: 886500-49-8
UN: 3265
ADR: 8,III