4-Methoxy-3-methyl-5-(trifluoromethyl)benzyl bromide 97%
Leverantör: Apollo Scientific
Fara
PC302351-1G PC302351-5G
APOSPC302351-1GEA 2300 SEK
APOSPC302351-1G APOSPC302351-5G
4-Methoxy-3-methyl-5-(trifluoromethyl)benzyl bromide 97%
4-Methoxy-3-methyl-5-(trifluoromethyl)benzyl bromide
Formel: C₁₀H₁₀BrF₃O
MW: 283,085 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD23795340
CAS nummer: 1431329-69-9
UN: 1759
ADR: 8,II