4-Ethoxy-2,3-difluorophenylacetonitrile
Leverantör: Apollo Scientific
PC302330-1G PC302330-5G
APOSPC302330-1GEA 1375 SEK
APOSPC302330-1G APOSPC302330-5G
4-Ethoxy-2,3-difluorophenylacetonitrile
4-Ethoxy-2,3-difluorophenylacetonitrile
Formel: C₁₀H₉F₂NO
MW: 197.181 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09258706
CAS nummer: 1017779-59-7