3-Bromo-5-(trifluoromethoxy)cinnamic acid 98%
Leverantör: Apollo Scientific
|
Varning
|
PC302283-1G
PC302283-5G
PC302283-10G
APOSPC302283-1GEA
1750
SEK
APOSPC302283-1G
APOSPC302283-5G
APOSPC302283-10G
3-Bromo-5-(trifluoromethoxy)cinnamic acid 98%
3-Bromo-5-(trifluoromethoxy)cinnamic acid
Formel:
C₁₀H₆BrF₃O₃ MW: 311,052 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD09999513 CAS nummer: 1092460-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...