3,6-Dichloro-2-fluorophenol 98%
Leverantör: Apollo Scientific
Varning
PC302275-1G PC302275-5G PC302275-10G
APOSPC302275-1GEA 1525 SEK
APOSPC302275-1G APOSPC302275-5G APOSPC302275-10G
3,6-Dichloro-2-fluorophenol 98%
3,6-Dichloro-2-fluorophenol
Formel: C₆H₃Cl₂FO
MW: 180,992 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09025387
CAS nummer: 916420-67-2