2-Fluoro-4,6-bis(trifluoromethyl)benzyl bromide 98%
Leverantör: Apollo Scientific
PC302242-1G PC302242-5G PC302242-10G
APOSPC302242-1GEA 1150 SEK
APOSPC302242-1G APOSPC302242-5G APOSPC302242-10G
2-Fluoro-4,6-bis(trifluoromethyl)benzyl bromide 98%
2-Fluoro-4,6-bis(trifluoromethyl)benzyl bromide
Formel: C₉H₄BrF₇
MW: 325.021 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09258673
CAS nummer: 1017778-46-9