4-Fluoro-2-methyl-DL-phenylalanine

Leverantör: Apollo Scientific

PC302148-1G PC302148-5G
APOSPC302148-1GEA 1400 SEK
APOSPC302148-1G APOSPC302148-5G
4-Fluoro-2-methyl-DL-phenylalanine
4-Fluoro-2-methyl-DL-phenylalanine
Formel: C₁₀H₁₂FNO₂
MW: 197,206 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04116030
CAS nummer: 1259993-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR