4-Ethoxy-2,3-difluorocinnamic acid 97%
Leverantör: Apollo Scientific
PC302108-1G PC302108-5G
APOSPC302108-1GEA 1100 SEK
APOSPC302108-1G APOSPC302108-5G
4-Ethoxy-2,3-difluorocinnamic acid 97%
4-Ethoxy-2,3-difluorocinnamic acid
Formel: C₁₁H₁₀F₂O₃
MW: 228,192 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09258709
CAS nummer: 1017779-66-6