2,3-Difluoro-4-methoxycinnamic acid
Leverantör: Apollo Scientific
PC302068-1G PC302068-5G
APOSPC302068-1GEA 1875 SEK
APOSPC302068-1G APOSPC302068-5G
2,3-Difluoro-4-methoxycinnamic acid
2,3-Difluoro-4-methoxycinnamic acid
Formel: C₁₀H₈F₂O₃
MW: 214.166 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06660222
CAS nummer: 886499-79-2