3-[3-Fluoro-5-(trifluoromethyl)phenyl]propionic acid 98%
Leverantör: Apollo Scientific
Varning
PC302043-1G PC302043-5G PC302043-10G
APOSPC302043-1GEA 1025 SEK
APOSPC302043-1G APOSPC302043-5G APOSPC302043-10G
3-[3-Fluoro-5-(trifluoromethyl)phenyl]propionic acid 98%
3-[3-Fluoro-5-(trifluoromethyl)phenyl]propionic acid
Formel: C₁₀H₈F₄O₂
MW: 236.163 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09025358
CAS nummer: 916420-41-2