4-(6-Bromo-8-fluoroimidazo[1,2-a]pyridin-2-yl)benzonitrile
Leverantör: Apollo Scientific
Varning
PC301015-1G PC301015-250MG
APOSPC301015-1GEA 6175 SEK
APOSPC301015-1G APOSPC301015-250MG
4-(6-Bromo-8-fluoroimidazo[1,2-a]pyridin-2-yl)benzonitrile
4-(6-Bromo-8-fluoroimidazo[1,2-a]pyridin-2-yl)benzonitrile
Formel: C₁₄H₇BrFN₃
MW: 316,128 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD32066751
CAS nummer: 2379918-56-4