Ethyl 2,4-difluoro-5-iodobenzoate
Leverantör: Apollo Scientific
Varning
PC300981-1G PC300981-5G
APOSPC300981-1GEA 410 SEK
APOSPC300981-1G APOSPC300981-5G
Ethyl 2,4-difluoro-5-iodobenzoate
Ethyl 2,4-difluoro-5-iodobenzoate
Formel: C₉H₇F₂IO₂
MW: 312.052 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD24107026
CAS nummer: 1541937-20-5