2-(2-Bromo-4-fluorophenoxy)-4-(trifluoromethyl)pyrimidine
Leverantör: Apollo Scientific
PC300969-10G PC300969-1G PC300969-5G
APOSPC300969-10GEA 18575 SEK
APOSPC300969-10G APOSPC300969-1G APOSPC300969-5G
2-(2-Bromo-4-fluorophenoxy)-4-(trifluoromethyl)pyrimidine
2-(2-Bromo-4-fluorophenoxy)-4-(trifluoromethyl)pyrimidine
Formel: C₁₁H₅BrF₄N₂O
MW: 337,068 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11520135
CAS nummer: 1455953-78-2