Ethyl (6-fluoroimidazo[1,2-a]pyridin-2-yl)acetate
Leverantör: Apollo Scientific
Varning
PC300896-1G PC300896-5G
APOSPC300896-1GEA 6050 SEK
APOSPC300896-1G APOSPC300896-5G
Ethyl (6-fluoroimidazo[1,2-a]pyridin-2-yl)acetate
Ethyl (6-fluoroimidazo[1,2-a]pyridin-2-yl)acetate
Formel: C₁₁H₁₁FN₂O₂
MW: 222.216 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD22552235
CAS nummer: 1315359-46-6