1,1,1-Trifluoro-3-[4-(4-nitrophenyl)piperazin-1-yl]propan-2-ol
Leverantör: Apollo Scientific
PC300882-1G PC300882-5G
APOSPC300882-1GEA 3325 SEK
APOSPC300882-1G APOSPC300882-5G
1,1,1-Trifluoro-3-[4-(4-nitrophenyl)piperazin-1-yl]propan-2-ol
1,1,1-Trifluoro-3-[4-(4-nitrophenyl)piperazin-1-yl]propan-2-ol
Formel: C₁₃H₁₆F₃N₃O₃
MW: 319.28 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02186261
CAS nummer: 478258-73-0