N'-(2,4-Dibromo-6-fluorophenyl)-N,N-diisopropylthiourea
Leverantör: Apollo Scientific
PC300860-1G PC300860-5G
APOSPC300860-1GEA 2150 SEK
APOSPC300860-1G APOSPC300860-5G
N'-(2,4-Dibromo-6-fluorophenyl)-N,N-diisopropylthiourea
N'-(2,4-Dibromo-6-fluorophenyl)-N,N-diisopropylthiourea
Formel: C₁₃H₁₇Br₂FN₂S
MW: 412,159 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28987634
CAS nummer: 1980054-14-5