N-(2,4-Dibromo-6-fluorophenyl)thiourea
Leverantör: Apollo Scientific
PC300824-1G PC300824-5G
APOSPC300824-1GEA 1725 SEK
APOSPC300824-1G APOSPC300824-5G
N-(2,4-Dibromo-6-fluorophenyl)thiourea
N-(2,4-Dibromo-6-fluorophenyl)thiourea
Formel: C₇H₅Br₂FN₂S
MW: 327,999 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28148245
CAS nummer: 1823494-62-7