Methyl 4-(2,6-dimethylmorpholin-4-yl)-2,3,5-trifluorobenzoate
Leverantör: Apollo Scientific
PC300782-1G PC300782-5G PC300782-250MG
APOSPC300782-1GEA 2750 SEK
APOSPC300782-1G APOSPC300782-5G APOSPC300782-250MG
Methyl 4-(2,6-dimethylmorpholin-4-yl)-2,3,5-trifluorobenzoate
Methyl 4-(2,6-dimethylmorpholin-4-yl)-2,3,5-trifluorobenzoate
Formel: C₁₄H₁₆F₃NO₃
Storage Temperature: Omgivande
MDL Number: MFCD28103344
CAS nummer: 1858251-69-0