Methyl 2,3,5-trifluoro-4-(4-methylpiperazin-1-yl)benzoate
Leverantör: Apollo Scientific
PC300764-1G PC300764-5G
APOSPC300764-1GEA 2925 SEK
APOSPC300764-1G APOSPC300764-5G
Methyl 2,3,5-trifluoro-4-(4-methylpiperazin-1-yl)benzoate
Methyl 2,3,5-trifluoro-4-(4-methylpiperazin-1-yl)benzoate
Formel: C₁₃H₁₅F₃N₂O₂
Storage Temperature: Omgivande
MDL Number: MFCD28040508
CAS nummer: 1858256-51-5