Methyl 5-chloro-2-fluorobenzoate
Leverantör: Apollo Scientific
Varning
PC300717-1G PC300717-5G PC300717-25G
APOSPC300717-1GEA 515 SEK
APOSPC300717-1G APOSPC300717-5G APOSPC300717-25G
Methyl 5-chloro-2-fluorobenzoate
Methyl 5-chloro-2-fluorobenzoate
Formel: C₈H₆ClFO₂
Storage Temperature: Omgivande
MDL Number: MFCD06204184
CAS nummer: 57381-36-9