4,4,4-Trifluorobutyl trifluoromethanesulphonate 97%

Leverantör: Apollo Scientific
Fara

PC2971-1G PC2971-5G PC2971-250MG
APOSPC2971-1GEA 2275 SEK
APOSPC2971-1G APOSPC2971-5G APOSPC2971-250MG
4,4,4-Trifluorobutyl trifluoromethanesulphonate 97%
4,4,4-Trifluorobutyl trifluoromethanesulphonate
Formel: C₅H₆F₆O₃S
MW: 260,155 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07784317
CAS nummer: 885275-65-0
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR