5-Fluoro-3-methylbenzo[b]thiophene-2-sulphonyl chloride
Leverantör: Apollo Scientific
Fara
Synonyms: 5-Fluoro-3-methyl-1-benzothiophene-2-sulphonyl chloride, 2-(Chlorosulphonyl)-5-fluoro-3-methylbenzo[b]thiophene
PC2907-1G PC2907-5G
APOSPC2907-1GEA 2075 SEK
APOSPC2907-1G APOSPC2907-5G
5-Fluoro-3-methylbenzo[b]thiophene-2-sulphonyl chloride
5-Fluoro-3-methylbenzo[b]thiophene-2-sulphonyl chloride
Formel: C₉H₆ClFO₂S₂
MW: 264,724 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD02091512
CAS nummer: 404964-34-7
UN: 3261
ADR: 8,III