8-Chloro-3-methyl-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Leverantör: Apollo Scientific
Varning
PC28304-1G PC28304-5G
APOSPC28304-1GEA 1550 SEK
APOSPC28304-1G APOSPC28304-5G
8-Chloro-3-methyl-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
8-Chloro-3-methyl-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Formel: C₈H₅ClF₃N₃
MW: 235,594 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12406874
CAS nummer: 1823182-33-7