4,5-Dichloro-2-(3-chloro-5-(trifluoromethyl)pyridin-2-yl)-6-methylpyrimidine
Leverantör: Apollo Scientific
Varning
PC28285-1G PC28285-250MG
APOSPC28285-1GEA 6175 SEK
APOSPC28285-1G APOSPC28285-250MG
4,5-Dichloro-2-(3-chloro-5-(trifluoromethyl)pyridin-2-yl)-6-methylpyrimidine
4,5-Dichloro-2-(3-chloro-5-(trifluoromethyl)pyridin-2-yl)-6-methylpyrimidine
Formel: C₁₁H₅Cl₃F₃N₃
MW: 342,532 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD31812110
CAS nummer: 2379918-50-8