1-(8-Chloro-3-(methylsulphonyl)-6-(trifluoromethyl)imidazo[1,5-a]pyridin-1-yl)-2,2,2-trifluoroethanone
Leverantör: Apollo Scientific
Varning
PC28274-1G PC28274-250MG
APOSPC28274-1GEA 1700 SEK
APOSPC28274-1G APOSPC28274-250MG
1-(8-Chloro-3-(methylsulphonyl)-6-(trifluoromethyl)imidazo[1,5-a]pyridin-1-yl)-2,2,2-trifluoroethanone
1-(8-Chloro-3-(methylsulphonyl)-6-(trifluoromethyl)imidazo[1,5-a]pyridin-1-yl)-2,2,2-trifluoroethanone
Formel: C₁₁H₅ClF₆N₂O₃S
MW: 394.677 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD31812112
CAS nummer: 2379918-52-0