3-Bromo-8-chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Leverantör: Apollo Scientific
Varning
PC28261-1G PC28261-250MG
APOSPC28261-1GEA 2575 SEK
APOSPC28261-1G APOSPC28261-250MG
3-Bromo-8-chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
3-Bromo-8-chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Formel: C₇H₂BrClF₃N₃
MW: 300.463 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12406873
CAS nummer: 1823182-57-5