4-Chloro-6-fluoro-8-(trifluoromethyl)quinoline
Leverantör: Apollo Scientific
Varning
PC28243-1G PC28243-250MG
APOSPC28243-1GEA 6175 SEK
APOSPC28243-1G APOSPC28243-250MG
4-Chloro-6-fluoro-8-(trifluoromethyl)quinoline
4-Chloro-6-fluoro-8-(trifluoromethyl)quinoline
Formel: C₁₀H₄ClF₄N
MW: 249,592 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28010394
CAS nummer: 1065093-26-6