2-Fluoro-4-(hydroxymethyl)benzoic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: (4-Carboxy-3-fluorophenyl)methanol, 4-Carboxy-3-fluorobenzyl alcohol
PC2809-1G PC2809-250MG PC2809-5G
APOSPC2809-1GEA 1700 SEK
APOSPC2809-1G APOSPC2809-250MG APOSPC2809-5G
2-Fluoro-4-(hydroxymethyl)benzoic acid 98%
2-Fluoro-4-(hydroxymethyl)benzoic acid
Formel: C₈H₇FO₃
MW: 170,138 g/mol
Smältpunkt: 170…175 °C
Storage Temperature: Omgivande
MDL Number: MFCD08741375
CAS nummer: 214554-18-4