2-Fluorodiphenylamine 95%

Leverantör: Apollo Scientific
Varning

PC2798-250MG PC2798-100MG
APOSPC2798-250MGEA 2150 SEK
APOSPC2798-250MG APOSPC2798-100MG
2-Fluorodiphenylamine 95%
2-Fluorodiphenylamine
Formel: C₁₂H₁₀FN
Kokpunkt: 147…148 °C (15 mm Hg)
Densitet: 1,165 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD03094200
CAS nummer: 328-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR