3-Fluoro-5-methoxybenzyl alcohol 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 3-Fluloro-5-methoxybenzyl alcohol, (3-Fluoro-5-methoxyphenyl)methanol, 3-Fluoro-5-(hydroxymethyl)anisole
PC2796-5G PC2796-1G PC2796-10G
APOSPC2796-5GEA 3525 SEK
APOSPC2796-5G APOSPC2796-1G APOSPC2796-10G
3-Fluoro-5-methoxybenzyl alcohol 98%
3-Fluoro-5-methoxybenzyl alcohol
Formel: C₈H₉FO₂
MW: 156,154 g/mol
Smältpunkt: 27…30 °C
Storage Temperature: Omgivande
MDL Number: MFCD08741377
CAS nummer: 914637-27-7