4-Bromo-2-fluoro-5-nitrobenzyl fluoride 95%
Leverantör: Apollo Scientific
Fara
PC27922-1G PC27922-250MG
APOSPC27922-1GEA 7175 SEK
APOSPC27922-1G APOSPC27922-250MG
4-Bromo-2-fluoro-5-nitrobenzyl fluoride 95%
4-Bromo-2-fluoro-5-nitrobenzyl fluoride
Formel: C₇H₄BrF₂NO₂
MW: 252.013 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28738758
CAS nummer: 1807004-77-8
UN: 3265
ADR: 8 II