3-Bromo-4,5-difluorobenzyl fluoride 95%
Leverantör: Apollo Scientific
|
Fara
|
PC27915-1G
PC27915-250MG
APOSPC27915-1GEA
7200
SEK
APOSPC27915-1G
APOSPC27915-250MG
3-Bromo-4,5-difluorobenzyl fluoride 95%
3-Bromo-4,5-difluorobenzyl fluoride
Formel:
C₇H₄BrF₃ MW: 225,006 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD31664902 CAS nummer: 1785082-71-4 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...