3-Fluoro-5-methoxyaniline 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Fluoro-m-anisidine, 3-Amino-5-fluoroanisole
PC2791-1G PC2791-5G PC2791-25G
APOSPC2791-1GEA 585 SEK
APOSPC2791-1G APOSPC2791-5G APOSPC2791-25G
3-Fluoro-5-methoxyaniline 98%
3-Fluoro-5-methoxyaniline
Formel: C₇H₈FNO
MW: 141,143 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00077535
CAS nummer: 2339-58-4