N-Fmoc-6-Fluoro-L-tryptophan
Leverantör: Apollo Scientific
Synonyms: N-Fmoc-6-fluoro-L-tryptophan, Fmoc-Trp(6-F)-OH
PC27891-100MG PC27891-250MG PC27891-50MG
APOSPC27891-100MGEA 3250 SEK
APOSPC27891-100MG APOSPC27891-250MG APOSPC27891-50MG
N-Fmoc-6-Fluoro-L-tryptophan
N-Fmoc-6-Fluoro-L-tryptophan
Formel: C₂₆H₂₁FN₂O₄
MW: 444,454 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD29917530
CAS nummer: 908847-01-8