4-Bromo-2-fluoro-6-nitrotoluene
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Bromo-3-fluoro-2-methylnitrobenzene
PC2784-1G PC2784-5G
APOSPC2784-1GEA 260 SEK
APOSPC2784-1G APOSPC2784-5G
4-Bromo-2-fluoro-6-nitrotoluene
4-Bromo-2-fluoro-6-nitrotoluene
Formel: C₇H₅BrFNO₂
MW: 234.023 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03094190
CAS nummer: 502496-34-6