4-Fluoro-3-(methylsulphonyl)benzonitrile 97%

Leverantör: Apollo Scientific
Varning

Synonyms: 5-Cyano-2-fluorophenyl methyl sulphone

PC2627-1G PC2627-5G
APOSPC2627-1GEA 1175 SEK
APOSPC2627-1G APOSPC2627-5G
4-Fluoro-3-(methylsulphonyl)benzonitrile 97%
4-Fluoro-3-(methylsulphonyl)benzonitrile
Formel: C₈H₆FNO₂S
MW: 199,202 g/mol
Smältpunkt: 128,9…130.2 °C
Storage Temperature: Omgivande
MDL Number: MFCD08458081
CAS nummer: 914636-47-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR