2-Chloro-4,5-difluorobenzoyl chloride 98%
Leverantör: Apollo Scientific
Fara
PC2598-1G PC2598-100G PC2598-25G
APOSPC2598-1GEA 175 SEK
APOSPC2598-1G APOSPC2598-100G APOSPC2598-25G
2-Chloro-4,5-difluorobenzoyl chloride 98%
2-Chloro-4,5-difluorobenzoyl chloride
Formel: C₇H₂Cl₂F₂O
MW: 210,993 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04972708
CAS nummer: 121872-95-5
UN: 3265
ADR: 8,III