4-Bromo-2-(methoxymethyl)fluorobenzene 96%
Leverantör: Apollo Scientific
Synonyms: 4-Bromo-1-fluoro-2-(methoxymethyl)benzene
PC2423-100MG PC2423-250MG PC2423-1G
APOSPC2423-100MGEA 1050 SEK
APOSPC2423-100MG APOSPC2423-250MG APOSPC2423-1G
4-Bromo-2-(methoxymethyl)fluorobenzene 96%
4-Bromo-2-(methoxymethyl)fluorobenzene
Formel: C₈H₈BrFO
MW: 219.051 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04116325
CAS nummer: 337535-43-0