Vinyl perfluorononanoate
Leverantör: Apollo Scientific
|
Varning
|
PC2387-1G
PC2387-5G
APOSPC2387-1GEA
1900
SEK
APOSPC2387-1G
APOSPC2387-5G
Vinyl perfluorononanoate
Vinyl perfluorononanoate
Formel:
C₁₁H₃F₁₇O₂ Storage Temperature: Omgivande |
MDL Number:
MFCD00078320 CAS nummer: 54640-64-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...