3-(Difluoromethyl)-1-methyl-1H-pyrazol-5(4H)-one ≥98%
Leverantör: Apollo Scientific
PC210128-100MG PC210128-1G PC210128-250MG
APOSPC210128-100MGEA 2950 SEK
APOSPC210128-100MG APOSPC210128-1G APOSPC210128-250MG
3-(Difluoromethyl)-1-methyl-1H-pyrazol-5(4H)-one ≥98%
3-(Difluoromethyl)-1-methyl-1H-pyrazol-5(4H)-one
Formel: C₅H₆F₂N₂O
MW: 148,111 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD17240576
CAS nummer: 1048018-91-2