4-(Heptafluoroisopropoxy)-1-iodooctafluorobutane

Leverantör: Apollo Scientific

PC2101-25G PC2101-5G
APOSPC2101-25GEA 8200 SEK
APOSPC2101-25G APOSPC2101-5G
4-(Heptafluoroisopropoxy)-1-iodooctafluorobutane
4-(Heptafluoroisopropoxy)-1-iodooctafluorobutane
Formel: C₇F₁₅IO
Kokpunkt: 40…43 °C (25 mm Hg)
Storage Temperature: Omgivande
MDL Number: MFCD09763606
CAS nummer: 25080-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR